Learn More
TARGETMOL CHEMICALS INC UCB-9260 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T139515MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. UCB-9260 inhibits TNF signaling by stabilizing an asymmetric form of the trimer. Purity 99.65%
Catalog No. 50-288-0099