Learn More
TARGETMOL CHEMICALS INC UCB7362 hydrochloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T69567L1MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. UCB7362 hydrochloride (UCB7362 HCl) is an orally active plasmepsin X (PMX) inhibitor (IC50 7 nM) with antimalarial activity that inhibits parasite growth. Purity 99.1%
Catalog No. 50-287-7902