Learn More
Medchemexpress LLC Uck2 inhibitor-1 | 902289-98-9 | 96.3% | 483.54 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14839450MG
Encompass_Preferred
UCK2 Inhibitor-1 (Compound 20874830-2) is a non-competitive UCK2 inhibitor with an IC50 of 4.7 μM. It acts as a non-competitive inhibitor against uridine and ATP, functionally inhibiting the enzymatic activity of UCK2. This inhibitor suppresses the uridine salvage pathway and can be utilized in chronic myeloid leukemia research.
  • Acts as a non-competitive UCK2 inhibitor
  • Inhibits enzymatic activity of UCK2
  • Suppresses the uridine salvage pathway
  • Useful in chronic myeloid leukemia research
  • Has an IC50 of 4.7 μM against UCK2
Catalog No. 50-003-60166