Learn More
Cayman Chemical UCLA gp130 2 50mg
SDP
Supplier:  Cayman Chemical 3555150 MG
Encompass_Preferred
A humanin peptide mimetic; inhibits NMDA-induced LDH release in primary mouse hippocampal neurons at 10 µM, an effect that can be reversed by SC144
Catalog No. 50-291-3616