Learn More
Selleck Chemical LLC UCPH-101 S0297-1G
SDP
Supplier:  Selleck Chemical LLC S02971G
Encompass_Preferred
UCPH-101 is an inhibitor of excitatory amino acid transporter subtype 1 (EAAT1) with IC50 of 0 66 M at EAAT1-HEK293 cells in the [3H]-D-Asp uptake assay
Catalog No. 50-000-42305