Learn More
Medchemexpress LLC UCPH-102 | 2165322-94-9 | MFCD32201047 | 98.0% | 422.21 g/mol | C17H16BrF4NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117370100MG
Encompass_Preferred
USP25/28 inhibitor AZ1 is a selective small-molecule inhibitor of the deubiquitinases USP25 and USP28, supplied as a high-purity research compound for biochemical, cellular, and in vivo studies.
  • Selective dual inhibitor of USP25 and USP28.
  • Orally active in in vivo models.
  • High purity suitable for research use (98.0%).
  • Suitable for biochemical assays and cell-based studies.
  • Available in multiple sizes, including 100 MG for dosing flexibility.
Catalog No. 50-002-26906