Learn More
American Radiolabeled Chemicals Inc UDP 5,6-3H 1LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 19141MC
Encompass_Preferred
Uridine 5AE-5,6-3H diphosphate tetrasodium salt (UDP) 1LC in ethanol:water (1:1), 1 mCi/ml, 30-60 Ci/mmol, M.W. 427.16, Shipped in Dry Ice, Exclusive
Catalog No. 50-353-009
Add to Cart
Request a Quote