Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UDP-D-GalNAlk disodium | 2370947-52-5 | 689.37 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY164903A1MG

Encompass_Preferred

UDP-D-GalNAlk disodium is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Biochemical reagent for life science research.
  • Suitable as a biological material or organic compound.
  • Store at -20°C, sealed, and away from moisture.
  • Soluble in H2O at 50 mg/mL, requiring ultrasonic.

Catalog No. 50-003-91593


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.