Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UDP-GlcNAc disodium | 91183-98-1 | 99.9% | 651.32 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11217450MG

Encompass_Preferred

UDP-GlcNAc disodium salt (UDP-α-D-N-Acetylglucosamine disodium salt) is a donor substrate of O-GlcNAc transferase (OGT). OGT is a nucleocytoplasmic glycosyltransferase assigned to the GT41 family. This enzyme modifies thousands of proteins by adding a unique N-acetylglucosamine residue onto acceptor substrates mainly confined within the cytosol and nucleus.

  • Target: OGT
  • Pathway: Neuronal signaling
  • Storage: -20°C, sealed, away from moisture; unstable in solutions
  • Solubility (in vitro): H2O: 125 mg/mL
  • Molecular formula: C17H25N3Na2O17P2
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-38093


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.