Learn More
Medchemexpress LLC UDP-glucosamine (disodium) | 1355005-51-4 | 99.4% | 609.28 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1397931MG
Encompass_Preferred
UDP-glucosamine (UDP-GlcNAc) disodium is a substrate for O-GlcNAc transferase, which facilitates the attachment of O-GlcNAc to proteins. O-GlcNAcase is responsible for removing O-GlcNAc from proteins. This compound is the final product of the hexosamine biosynthesis pathway, primarily controlled by glucose-6-phosphate-Glutamine:fructose-6-phosphate amidotransferase (GFAT).
  • Substrate for O-GlcNAc transferase
  • End product of the hexosamine biosynthesis pathway
Catalog No. 50-003-51262