Learn More
WELCH MATERIALS INC Ultisil UHPLC HILIC, 1.8µm, 2.1×50mm W-PORT
Supplier:  WELCH MATERIALS INC H0020011210
Encompass_Preferred
Ultisil UHPLC HILIC, 1.8μm, 2.1×50mm, W-Port, pH range: 2.0-8.0, Pore Size: 120Å, USP listing: L3, USP listing: L3, , Surface Area: 320m2/g, endcapped.
Catalog No. NC2052563