Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O=C([C@]1(CO1)CO[Si](C)(C)C(C)(C)C)[C@H](CC(C)C)NC([C@H](C)NC(CCCCCC)=O)=O | 1000313-40-5 | 99.6% | 484.74 | C25H48N2O5Si | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1190375MG

Encompass_Preferred

UK-101 is a selective immunoproteasome β1i (LMP2) inhibitor for laboratory research. It shows reported potency with an IC50 of about 104 nM and is supplied as a high-purity solid suitable for in vitro studies; recommended solvent is DMSO.

  • Selective immunoproteasome β1i (LMP2) inhibitor with IC50 ≈104 nM.
  • High purity (≈99.6%).
  • Available in small research quantities, e.g., 5 mg.
  • Recommended solvent: DMSO for dissolution.
  • Powder storage: -20°C (3 years) or 4°C (2 years); solutions: -80°C (6 months).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-29462


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.