Learn More
TARGETMOL CHEMICALS INC UK-101 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T132475MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. UK-101 is a potent and selective inhibitor of the immunoproteasome LMP2 inhibiting (beta)1i (LMP2) (beta)1c (LMP2) and (beta)5 (LMP2) with IC50s of 104 nM 15 uM and 1 uM. UK-101 has a 144-fold and 10-fold higher affinity for (beta)1i than for the distribution of (beta)1c and (beta)5 subunits. UK-101 induces apoptosis and can be used to study prostate cancer related diseases. purity: 99%
Catalog No. 50-258-1366