Learn More
Cayman Chemical UK 122trIfluoROCTAte salt 10mg
SDP
Supplier:  Cayman Chemical 900121810 MG
Encompass_Preferred
A uPA inhibitor (IC50 = 0.2 µM); selective for uPA over tPA, trypsin, plasmin, and thrombin (IC50s = >100 µM for all); inhibits the migration and invasion of CFPAC-1 cells at 100 µM; prevents glutamate-induced phase shifts in isolated suprachiasmatic nucleus slices from tPA-/- but not wild-type mice at 0.2 µM; decreases disease severity and colorectal levels of RANTES in a mouse model of DSS-induced colitis at 4 mg/kg
Catalog No. 50-289-2758