Learn More
Medchemexpress LLC Brimonidine (tartrate) | 70359-46-5 | 99.7% | 100 MG

Supplier: Medchemexpress LLC HYB0659A100MG
Brimonidine tartrate is a full α2-adrenergic receptor (α2-AR) agonist, primarily used for research purposes. It is a solid, white to off-white compound with high purity.
- Full α2-adrenergic receptor (α2-AR) agonist
- High purity of 99.7%
- Solid appearance, white to off-white color
- CAS No. 70359-46-5
- Molecular weight 442.22
- Molecular formula C15H16BrN5O6
- Melting point 207-208°C
- Target: Adrenergic Receptor
- Pathway: GPCR/G Protein; Neuronal Signaling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.