Learn More
Medchemexpress LLC HY-101214 5mg Medchemexpress, UK-371804 CAS:256477-09-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012145MG
Encompass_Preferred
Medchemexpress, HY-101214 5mg UK-371804 CAS:256477-09-5 UK-371804 is a urokinase-type plasminogen activator ( uPA ) inhibitor with a K i of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2067