Learn More
Selleck Chemical LLC UK 5099, 100mg, Purity: 99.99%
SDP
Supplier:  Selleck Chemical LLC S5317100MG
Encompass_Preferred
UK5099 (PF-1005023) is a potent inhibitor of the mitochondrial pyruvate carrier, inhibiting pyruvate transport across the plasma membrane of trypanosomes with Ki value of 49 μM.
Catalog No. NC2881483