Learn More
TARGETMOL CHEMICALS INC UK-5099 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T444125MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UK-5099 (PF-1005023) is a specific and potent inhibitor of MPC carrier activity [1]. It potently inhibits pyruvate-dependent oxygen consumption by rat heart mitochondria (IC50 50 nM) [1]. purity: 100%
Catalog No. 50-257-9883