Learn More
Medchemexpress LLC UK4b | 2171000-87-4 | 96.17% | 404.89 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1623875MG
Encompass_Preferred
UK4b is a highly selective microsomal prostaglandin E2 synthase-1 (mPGES-1) inhibitor. It possesses anti-inflammatory and analgesic effects. UK4b has also been shown to block the growth of abdominal aortic aneurysms in mice.
  • Highly selective microsomal prostaglandin E2 synthase-1 (mPGES-1) inhibitor.
  • Possesses anti-inflammatory effects.
  • Possesses analgesic effects.
  • Can block the growth of abdominal aortic aneurysms in mice.
Catalog No. 50-003-92763