Learn More
ABclonal Technology ULBP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2116120UL
Encompass_Preferred
The protein encoded by this gene is a ligand of natural killer group 2 member D (NKG2D) an immune system-activating receptor on NK cells and T-cells Binding of the encoded ligand to NKG2D leads to activation of several signal transduction pathways including those of JAK2 STAT5 ERK and PI3K kinase Akt Also in cytomegalovirus-infected cells this ligand binds the UL16 glycoprotein and is prevented from activating the immune system Three transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-07507