Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD54
Supplier:  Biolegend 353133
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD54 [HA58]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC, IHC-F; Size: 100 ug
Catalog No. 50-207-7210