Learn More
Biolegend Ultra-LEAF™ Purified anti-human/mouse CD49f
Supplier:  Biolegend 313637
Encompass_Preferred
Ultra-LEAF™ Purified anti-human/mouse CD49f [GoH3]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Mouse, Baboon, Capuchin Monkey, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Rhesus, Sheep (Ovine), Swine (Pig, Porcine); Apps: FC, ICC, IHC-F, IP, Block; Size: 100 ug
Catalog No. 50-207-3658