Learn More
Biolegend Ultra-LEAF™ Purified anti-human TSLP
Supplier:  Biolegend 512206
Encompass_Preferred
Ultra-LEAF™ Purified anti-human TSLP [15B11.3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, Neut; Size: 1 mg
Catalog No. 50-208-5833