Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IFN-γ
Supplier:  Biolegend 505848
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, ELISPOT Capture, CyTOF, Neut, ICFC, IHC, WB; Size: 25 mg
Catalog No. 50-208-5712