Learn More
Medchemexpress LLC Ulefnersen sodium | 2589926-27-0 | 98.47% | 7060 (free acid) | 1 MG
SDP
Supplier:  Medchemexpress LLC HY147410A1MG
Encompass_Preferred
Ulefnersen sodium (ION363) is an Antisense Oligonucleotide (ASO) that targets the 6th intron of the fused-in sarcoma (FUS) transcript to silence FUS in a non-allele-specific manner.
  • Reduces postnatal levels of FUS protein in the brain and spinal cord
  • Delays motor neuron degeneration in disease-relevant mouse models of ALS-FUS
  • Used in the research of Amyotrophic Lateral Sclerosis (ALS)
Catalog No. 50-003-59669