Learn More
Vector Laboratories Ulex Europaeus Agglutinin I (UEA I) Biotinylated 2 mg
SDP
Supplier:  Vector Laboratories B10652
Encompass
Ulex europaeus agglutinin I (UAE I) binds to many glycoproteins and glycolipids containing α-linked fucose residues, such as ABO blood group glycoconjugates. This lectin preferentially binds blood group O cells and has been used to determine secretor status. It is also an established marker for human endothelial cells. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC9936752