Learn More
Medchemexpress LLC HY-14959 10mg Medchemexpress, Ulipristal CAS:159811-51-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1495910MG
Encompass_Preferred
Medchemexpress, HY-14959 10mg Ulipristal CAS:159811-51-5 Ulipristal (CDB 3236) is a selective progesterone receptor modulator (SPRM). Ulipristal binds to the progesteron receptor, thereby inhibiting PR-mediated gene expression, and interfering with progesterone activity in the reproductive system. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0571
 Restricted Item