Learn More
Selleck Chemical LLC Ulipristal E4845-100MG
SDP
Supplier:  Selleck Chemical LLC E4845100MG
Encompass_Preferred
Ulipristal(Deacetyl CDB-2914 CDB-3236) is a selective modulator of progesterone receptors used in a single dose as an emergency postcoital contraceptive
Catalog No. 50-000-42499