Learn More
Selleck Chemical LLC Ulixertinib
SDP
Supplier:  Selleck Chemical LLC S7854100MG
Encompass_Preferred
Ulixertinib (BVD-523 VRT752271) is a potent and reversible ERK1/ERK2 inhibitor with IC50 of 0 3 nM for ERK2 Phase 1
Catalog No. 50-000-00056