Learn More
Selleck Chemical LLC Ulixertinib (BVD-523) 25mg 869886-67-9 VRT752271
SDP
Supplier:  Selleck Chemical LLC S785425MG
Encompass_Preferred
Ulixertinib (BVD-523, VRT752271) is a potent and reversible ERK1/ERK2 inhibitor with IC50 of <0.3 nM for ERK2. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1839948