Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ULK-101 25mg | 2443816-45-1 | 460.45 | C22H16F4N4OS | 25 MG
SDP

Supplier:  Medchemexpress LLC HY11449025MG

Encompass_Preferred

ULK-101 is a potent, selective small-molecule inhibitor of ULK1 used in cellular and biochemical research to suppress autophagy and sensitize cancer cells to nutrient stress.

  • Potent ULK1 inhibition (IC50 = 1.6 nM) with lower activity on ULK2 (IC50 = 30 nM).
  • High purity suitable for research applications (99.91%).
  • Available as a powder in multiple pack sizes, including 25 MG, and as DMSO solution formats.
  • Storage-stable when kept at recommended temperatures for powder and solutions.
  • Molecular weight 460.45 and molecular formula C22H16F4N4OS.

Catalog No. 50-002-29726


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.