Learn More
Medchemexpress LLC ULK-101 50mg | 2443816-45-1 | 460.45 g/mol | C22H16F4N4OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11449050MG
Encompass_Preferred
ULK-101 is a potent, selective small-molecule inhibitor of Unc-51-like autophagy-activating kinase 1 (ULK1) that suppresses autophagy and sensitizes cancer cells to nutrient stress. It is characterized in vitro with low-nanomolar activity against ULK1 and shows activity versus ULK2. Supplied as a powder for laboratory research use only.
Catalog No. 50-002-29977