Learn More
TARGETMOL CHEMICALS INC ULK1-IN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T633435MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ULK1-IN-2 is a potent ULK1 inhibitor with potential anticancer activity inducing apoptosis while blocking autophagy.ULK1-IN-2 has high cytotoxicity against cancer cells with an IC50 value of 1.94 uM against A549 cells.ULK1-IN-2 can be used for the study of non-small-cell lung cancer. purity: 99%
Catalog No. 50-257-6528