Learn More
Medchemexpress LLC Ulotaront hydrochloride | 1310422-41-3 | 99.9% | 219.73 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHY136109
Encompass_Preferred
Ulotaront hydrochloride (SEP-363856 hydrochloride) is an orally active TAAR1 and 5-HT1A agonist and a CNS active psychotropic agent. It possesses a unique, non-D2/5-HT2A mechanism of action and exerts antipsychotic-like effects. It has potential for the study of schizophrenia.
  • Orally active TAAR1 and 5-HT1A agonist.
  • CNS active psychotropic agent.
  • Unique, non-D2/5-HT2A mechanism of action.
  • Exerts antipsychotic-like effects.
  • Potential for the study of schizophrenia.
  • High purity: 99.89%.
  • Available in solid and solution forms.
Catalog No. 50-003-64534