Learn More
Medchemexpress LLC Ulotaront hydrochloride | 1310422-41-3 | 99.9% | 219.73 g/mol | C9H14ClNOS | 5 MG

Supplier: Medchemexpress LLC HY1361095MG
Ulotaront hydrochloride is a research-stage TAAR1/5-HT receptor-active small molecule provided as the hydrochloride salt for preclinical studies. It is supplied in small quantities for laboratory research and is accompanied by datasheet and safety documentation.
- Purity 99.9%.
- Molecular formula C9H14ClNOS.
- Molecular weight 219.73 g/mol.
- Provided as hydrochloride salt for research use only.
- Supplied in small research pack sizes, including 5 mg.
- Datasheet and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.