Learn More
Sigma Aldrich Fine Chemicals Biosciences Ultima GoldTM LSC Cocktail
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L82862X5L
Encompass
Multi-purpose liquid scintillation cocktail (LSC) for a wide range of aqueous and non-aqueous samples.LSC will not diffuse through polyethylene vials. It provides superior detection efficiency for samples which exhibit severe quench in conventional cocktails and is biodegradable.
Catalog No. 50-291-4703