Learn More
WELCH MATERIALS INC Ultisil MM C18/SCX, 5μm, 4.6×150mm
Supplier:  WELCH MATERIALS INC H0023531041
Encompass_Preferred
Ultisil MM C18/SCX column features a mixed-mode stationary phase with C18 and SCX bonding, offering distinct selectivity from traditional single bonded phase columns. Featuring dual retention mechanisms of hydrophobic interaction and ion exchange, it is ideal for analyzing unknown compounds, particularly in metabolite research. It allows for the separation of highly polar compounds without the need for ion-pairing reagents, and allows for direct MS analysis.
Catalog No. NC3779944