Learn More
WELCH MATERIALS INC Ultisil XB-C18, 5μm, 4.6×150mm
Supplier:  WELCH MATERIALS INC H0020131041
Encompass_Preferred
Ultisil XB-C18 is one of the most versatile C18 analytical columns on the market. It features advanced bonding and double-endcapping technology, delivering nearly perfect peak shapes. This column is ideal for analyzing complex matrix samples and provides excellent peak shapes for acidic and basic compounds. It is particularly effective for the separation of traditional Chinese medicine compounds.
Catalog No. NC3779981