Learn More
WELCH MATERIALS INC Ultisil XB-C18, Cartridge: 5µm, 120Å, 4.6×10mm, 2pcs
Supplier:  WELCH MATERIALS INC H0080804001
Encompass_Preferred
Welch guard cartridges use the same packing materials as the analytical column to ensure the best analysis results. When HPLC column ID<4.0mm, it is recommended to use a 2.1×10mm cartridge, while when column ID≥4.0mm, a 4.6×10mm cartridge is recommended. 2.1×10mm cartridge holder: 00808-01107, 4.6×10mm cartridge holder: 00808-01101 or 00808-01108(direct connect).
Catalog No. NC3779950