Learn More
Bioworld Ultra Amplifying IF Kit: Cellular Stress- 150 slides -Eclipse, 1 Kit
Supplier:  Bioworld 217600131
Encompass_Preferred
The Ultra Amplifying IF Kit for the Detection of Cell Stress (Antibody Amplifier Eclipse version) contains a classic Antibody Amplifier, 1 500ml bottle of Amplifying Dilution Buffer, 1 500ml bottle of 20X Amplifying Wash Buffer, and 1 250ml bottle of 10X Epitope Unmasking Solution. This cell stress focus kit also includes 4 antibodies which are important indicators of cellular stress response: anti-DNA Damage, anti-Nitrotyrosine, anti-p53, and anti-p53-phospho-serine 15. These This kit provides enough reagents for processing 150 slides.
Catalog No. 50-199-085