Learn More
Perkin Elmer US LLC Ultra Clean Brass Triple Filter Kit, 0.125 in
Supplier:  Perkin Elmer US LLC N9306829
Encompass_Preferred
The triple combination filter kit is ideal for purifying GC/MS carrier gases. It contains oxygen, moisture and hydrocarbon scrubbers in one easy to change economical cartridge. There is of a range of brass and stainless steel base plate fittings and options for hydrocarbon trap to be helium or hydrogen specific, in addition to the general carrier gas.
Catalog No. 50-210-5217