Learn More
Biolegend Ultra-LEAF™ Purified anti-human/mouse/rat MR1
Supplier:  Biolegend 361109
Encompass_Preferred
Ultra-LEAF™ Purified anti-human/mouse/rat MR1 [26.5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat, Cross-Reactivity: Bovine; Apps: FC, ELISA, IP, Block; Size: 100 ug
Catalog No. NC1911538