Learn More
Biolegend Ultra-LEAFTM Purified anti-hum
Supplier:  Biolegend 629754
Encompass_Preferred
Ultra-LEAF™ Purified anti-human Ephrin-A1 [A16015A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: Neut; Size: 1 mg
Catalog No. 50-290-6400