Learn More
Rpc Ultra-Low™ Total Chlorine Test Strips, 0 - 0.2 ppm, 100/pk
Supplier: Rpc K1000118
RPC's Ultra-Low Total Chlorine Test Strips were designed and developed specifically for testing very low levels of chlorine and chloramines (TC) in water. Each test strip is individually foil wrapped and has a patented flow-through aperture. These features assure strip stability and maximum sensitivity. Seven color blocks with yellow-to-blue color shades make it easy to differentiate between values. RPC's Ultra-Low Total Chlorine Test Strips provide easy testing for ppm values of 0.0, 0.01, 0.02, 0.05, 0.1, 0.15, and 0.2. Use of a special, non-DPD reagent eliminates manganese interference that has caused problems for DPD tests. Use RPC's Ultra-Low Total Chlorine test strips to be confident that chlorine levels in your water do not exceed maximum levels called out in your procedures.