Learn More
Ted Pella Inc Ultra-Thin Blade, Single Edge, box of 100
SDP
Supplier:  Ted Pella Inc 12119
Encompass
Single edge, stainless steel blade. Ultra-thin, sharp edge blade with a cutting tip angle of 22° for use in vibratory slicers such as Microslicer™, Vibratome and others. Sold in a pack of 100.
Catalog No. NC1117893