Learn More
Waters Corp Ultrahydrogel Column, 250 Å, 6 μm, 7.8 mm x 30 cm
Supplier:  Waters Corp WAT011525
Encompass_Preferred
This Ultrahydrogel Column is a sophisticated aqueous-compatible gel column designed for analytical and preparative separations of water-soluble polymers. The gel is a cross-linked hydroxylated polymer with some residual carboxyl functionality. Ultrahydrogel DP contains some residual amine functionality. These columns are typically used to characterize compounds such as oligomers and biological substances like polysaccharides, nucleic acids, proteins, and peptides.
  • High resolution
  • Low adsorption
  • Wide range of molecular weight separations
  • Stability in a pH range of 2 to 12
  • Temperature range of 10°C to 80°C
Catalog No. 50-785-001