Learn More
Cayman Chemical UltraPure Water, 4L
SDP
Supplier:  Cayman Chemical 4000004
Encompass_Preferred
Water used for the preparation of EIA reagents and buffers. Deionized and free of organic contaminants. It has been purified by reverse osmosis followed by ion exchange and organic savenging.
Catalog No. NC9543422
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.