Learn More
Alpco Ltd ALPCO Ultrasensitive Insulin ELISA Jumbo Kit, Sandwich ELISA, FDA Class I, 10 plates
SDP
Supplier:  Alpco Ltd 80INSHUUE10
Encompass
The ALPCO Ultrasensitive Insulin ELISA Jumbo kit is designed for precise quantitative measurement of human insulin in serum or plasma samples. Its dual-monoclonal antibody format and high sensitivity of 0.135 µIU/mL distinguish it in clinical and research environments. With the capacity for processing up to 400 samples in duplicate and its FDA Class I device status for in vitro diagnostics, it ensures reliable and efficient results in insulin testing.
Catalog No. NC2235431