Learn More
Mercodia Inc ULTRASENSITIVE INSULIN ELISA
SDP
Supplier:  Mercodia Inc 10113201
Encompass_Preferred
A high quality enzyme immunoassay for specific quantification of human insulin in serum, plasma, or cell culture media. The Mercodia insulin assays have little or no cross-reactivity to insulin analogs, rat or mouse insulin, which enables distinguishing endogenous human insulin, both in patients undergoing treatment with insulin analogs, and in rat or mouse models. High sensitivity; Detection Limit 0.07 mU/L.
Catalog No. NC9919398