Learn More
Waters Corp Ultrastyragel Column, 1000 Å, 5 μm, 19 mm x 300 mm, Toluene
Supplier: Waters Corp WAT025868
This column is designed for high resolution preparative GPC applications. It contains hazardous material which may require special freight handling and additional charges.
- GPC chemistry
- SEC/GPC separation mode
- Polymer particle substrate
- pH range: 2 to 12 pH
- Molecular weight range: 200 to 30000
- Spherical particle shape
- 5 μm particle size
- 1000 Å pore size
- Column format
- Toluene packing solvent
- HPLC system
- SEC technique
- 19 mm inner diameter
- 300 mm length
- 1 pk units per package
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.